ZMQSFTSA1
Foot Pedal
Hands-free dispensing accessory for Milli-Q® IQ/IX, Milli-Q® EQ 7008/16 systems, and the Milli-Q IQ Element purification unit.
Synonym(s):
Milli-Q® Foot Pedal
About This Item
Recommended Products
packaging
pkg of 1 unit
manufacturer/tradename
E-POD®
Q-POD®
cable L
3 m
compatibility
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IQ Element
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7000
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Ideal for scientists who need free hands when accessing purified water.
- Supports lab sanitary protocols as users do not need to touch the system to access water.
- When combined with Volumetric Dispense mode, one press on the pedal starts the production of purified water at the point of dispense. Dispensing automatically stops at the set volume.
- Saves time when repeat dispensing the same volume.
- A 5 m cable provides large flexibility to position the pedal according to your lab design constraints.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service