Skip to Content
Merck
All Photos(1)

Documents

ZLXLDEGK2

Milli-Q

Degasser Upgrade Kit

For use with Milli-Q® HX and CLX series water purification systems

Synonym(s):

Degasser Upgrade Kit (Milli-Q® HX and CLX series)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
NACRES:
JA.13

packaging

pkg of 1 unit

compatibility

for use with AFS®
for use with Elix®
for use with Milli-Q® CLX
for use with Milli-Q® HX

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service