Skip to Content
Merck
All Photos(1)

Key Documents

XX2504758

Millipore

6-Place Sterility Test Manifold

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 182,700.00

CZK 182,700.00


Estimated to ship onSeptember 08, 2025Details


Request more information

Select a Size

Change View
1 EA
CZK 182,700.00

About This Item

UNSPSC Code:
41116129
eCl@ss:
32014001
NACRES:
NB.24

CZK 182,700.00


Estimated to ship onSeptember 08, 2025Details


Request more information

material

stainless steel body

Sterilization Compatibility

autoclavable compatible

input

sample type bottled water
sample type beer
sample type cosmetics
sample type pharmaceutical(s)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
sterility testing
water monitoring

shipped in

ambient

General description

Stainless steel manifolds for sterility testing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service