Skip to Content
Merck
All Photos(1)

Key Documents

XX2004708

Millipore

Millipore Stainless Steel Support Screen for Vacuum Filtration

47 mm

Synonym(s):

Stainless Steel Filter Screen

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 7,070.00

CZK 7,070.00


Estimated to ship onApril 16, 2025Details



Select a Size

Change View
1 EA
CZK 7,070.00

About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

CZK 7,070.00


Estimated to ship onApril 16, 2025Details


material

stainless steel screen

Quality Level

diam.

47 mm

shipped in

ambient

General description

Millipore Glass Vacuum Filtration Accessory Product
Stainless Steel Support Screen, 47 mm

These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.

This item is an accessory part for the Millipore 47 mm Classic Glass Filter Holder Kits and Millipore 47 mm All-Glass Filter Holder Kits.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the pore size?

    1 answer
    1. The pore size of this screen is 125 uM

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service