Skip to Content
Merck
All Photos(1)

Key Documents

XKEM00107

Millipore

Hand Vacuum Pump, polypropylene

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 2,410.00

CZK 2,410.00


Estimated to ship onApril 08, 2025Details



Select a Size

Change View
1 EA
CZK 2,410.00

About This Item

UNSPSC Code:
41105101
eCl@ss:
32012303
NACRES:
NB.24

CZK 2,410.00


Estimated to ship onApril 08, 2025Details


material

polypropylene

Quality Level

shipped in

ambient

General description

Use polypropylene syringes to dispense liquids into small containers and pressure-filter small volumes through filter holders of filter units with female Luer-Slip inlets. Syringes have male Luer-Slip outlets. An optional two-way vacuum valve, nylon male-to-male Luer adapter and rubber tubing with male Luer inlet converts this standard syringe into a hand vacuum pump. For sterile applications, an autoclavable stainless steel vacuum syringe with stainless steel, two-way valve is available.

Applications:
Small-Volume Liquid Filtration, Hand Pump for In-Field Filtration with Sterifil Units and Other Filter Holders

Principle

Vacuum

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hello, Could you please provide the diameter and volume of the outlet tube of this vacuum syringe. Thanks, Aman

    1 answer
    1. This product contains a surgical Latex rubber tubbing 14 inches long with an inner diameter of 5/32'' and a wall of 3/64''.

      Product XKEM00107 Hand Pump Assay contains a syringe: “XX1105005”. This is an “Off the Shelf” product that has a Luer-style tip. It meets ISO 594/1. It is also designed to be used with 0.312” (8mm) diameter tubing.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service