Skip to Content
Merck

VPPS102NB1

Millipore

Viresolve® Pro Shield

Virus prefilter, Viresolve® Pro Shield, Modus 1.2 format, filtration area 0.07 m²

Synonym(s):

Viresolve® Pro Shield Modus 1.2

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 30,500.00

CZK 30,500.00


Estimated to ship onJune 02, 2025Details



Select a Size

Change View
1 EA
CZK 30,500.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

CZK 30,500.00


Estimated to ship onJune 02, 2025Details


material

polyethersulfone

Quality Level

product line

EMPROVE® Filter

parameter

60 psig max. inlet pressure

device size

1.2 in.

impurities

<0.25 EU/mL USP bacterial endotoxins (LAL test, sample aqueous extraction)

General description

The Viresolve® Pro Solution provides a comprehensive, flexible template solution for viral clearance within mAb and recombinant protein bioproduction. This proven viral clearance solution delivers the highest levels of retention assurance and productivity across a broad range of feed stream characteristics.

Viresolve Pro Modus Devices are typically used for filtration of pilot to mid-scale batch volumes of proteins. The Viresolve® Pro Shield Modus 1.2 offers a filtration area of 0.07 m2 and has a primary use in pilot/small-volume processing.

Features and Benefits

  • Pilot and mid scale manufacturing
  • Three different size formats
  • Fully self contained (hardware holder not required)
  • Fully disposable single-use fluid path
  • Devices 100% tested with air/water diffusion and Binary Gas Test
  • Shield / Shield H 100% aerosol tested

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Preparation Note

This product was manufactured with materials that meet the criteria for a "non-fiber releasing" filter as defined in 21 CFR 210.3 (b) (6).

Analysis Note

TOC/Conductivity
After a controlled water flush of 100 L/m², samples exhibited less than 500 ppb TOC per USP <64> and less than 1.3 µS/cm per USP <64>.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
VIRESOLVE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

EMD Millipore Corporation certifies that this product complies with the European Pressure Equipment Directive, 97/23/EC of 29 May 1997. This product has been classified under Article 3 § 3 of the Pressure Vessel Directive. It has been designed and manufactured in accordance with sound engineering practice to ensure safe use. In compliance with Article 3 § 3 of this Pressure Vessel Directive, this product does not bear the CE mark.

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service