Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 filters | Estimated to ship on30 September 2026fromAreál Kühne+Nagel spol. s r.o. | 7 750,00 CZK |
About This Item
7 750,00 CZK
Estimated to ship on30 September 2026Details
Quality Segment
material
mixed cellulose esters (MCE) membrane, plain filter, white filter
description
90 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 25 discs
feature
hydrophilic
manufacturer/tradename
MF-Millipore™, Millipore
parameter
0.4 L/min-cm2 air flow rate, 1.5 mL/min-cm2 water flow rate, 75 °C max. temp.
diam.
90 mm
filter diam.
90 mm
thickness
100 μm
gravimetric extractables
1.5%
refractive index
n
matrix
MF-Millipore™
pore size
0.1 μm pore size, 74 % porosity
capacity
150 μg/cm2 binding capacity (protein)
bubble point
≥14.1 bar, air with water at 23 °C
shipped in
ambient
General description
MF-Millipore
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Application
Other Notes
- Organism Retention: Microorganism
- Mode of Action: Filtration (size exclusion)
- Application: General laboratory filtration
- Intended Use: Retention or removal of biological contaminants
- Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
- Storage Statement: Store in dry location away from heat source
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description MF-Millipore™, filter diam. 90 mm, hydrophilic | description MF-Millipore™, filter diam. 47 mm, hydrophilic | description MF-Millipore™, filter diam. 142 mm, hydrophilic | description MF-Millipore™, filter diam. 25 mm, hydrophilic |
| diam. 90 mm | diam. 47 mm | diam. 142 mm | diam. 25 mm |
| pore size 0.1 μm pore size, 74 % porosity | pore size 0.1 μm pore size, 74 % porosity | pore size 0.1 μm pore size, 74 % porosity | pore size 0.1 μm pore size, 74 % porosity |
| parameter 0.4 L/min-cm2 air flow rate, 75 °C max. temp., 1.5 mL/min-cm2 water flow rate | parameter 0.4 L/min-cm2 air flow rate, 1.5 mL/min-cm2 water flow rate, 75 °C max. temp. | parameter 0.4 L/min-cm2 air flow rate, 1.5 mL/min-cm2 water flow rate, 75 °C max. temp. | parameter 0.4 L/min-cm2 air flow rate, 1.5 mL/min-cm2 water flow rate, 75 °C max. temp. |
| manufacturer/tradename MF-Millipore™ | manufacturer/tradename MF-Millipore™ | manufacturer/tradename MF-Millipore™ | manufacturer/tradename MF-Millipore™ |
| material mixed cellulose esters (MCE) membrane, white filter, plain filter | material mixed cellulose esters (MCE) membrane, plain filter, white filter | material mixed cellulose esters (MCE) membrane, plain filter, white filter | material mixed cellulose esters (MCE) membrane, plain filter, white filter |
| feature hydrophilic | feature hydrophilic | feature hydrophilic | feature hydrophilic |
Still not finding the right product?
Explore all of our products under MCE Membrane Filter, 0.1 μm Pore Size
signalword
Danger
Storage Class
4.1B - Flammable solid hazardous materials
hcodes
Hazard Classifications
Aquatic Chronic 3 - ED ENV 1 - Flam. Sol. 1
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
9036-19-5
9036-19-5
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

