Skip to Content
Merck
All Photos(1)

Key Documents

USBA0250LA2S1

Millipore

Mobius®, Silver, Ultimus® Bin Assembly

Collapsible bin, side-ported with SterilEnz adapter and clamp, 250L

Synonym(s):

Mobius® Silver Ultimus Bin Assembly, Collapsible bin, side-ported with SterilEnz adapter and clamp, 250L

Sign Into View Organizational & Contract Pricing

Select a Size

USBA0250LA2S1
CZK 15,900.00

CZK 15,900.00


Estimated to ship onApril 14, 2025Details


Request more information

Select a Size

Change View
USBA0250LA2S1
CZK 15,900.00

About This Item

UNSPSC Code:
41123312

CZK 15,900.00


Estimated to ship onApril 14, 2025Details


Request more information

material

(Ultimus® Film)
C-Flex
polypropylene
silicone

Quality Level

Agency

meets EN 2.6.14
meets ISO 10993-4
meets JP 4.01
meets USP 661
meets USP 788
meets USP 85
meets USP 87
meets USP 88

sterility

sterile; γ-irradiated (at 25-40 kGy)

product line

Mobius®

parameter

2—60 °C temp. range (35.6—140 °F)

capacity

250 L

impurities

<0.25 EU/mL Aqueous extraction (LAL test)

application(s)

pharma/biopharma processes

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Our Mobius® Ultimus® Bin Assembly is a single-use assembly designed with our new Ultimus® film.
Ultimus® film was designed for more challenging single-use applications such as large-volume liquid processing. Our Ultimus® film technology provides enhanced bag strength, improved durability and leak resistance through a novel strength layer reinforced by woven nylon.
Our Ultimus® Film is now available in our Mobius® 3D assemblies of 100L, 200L, 250L, 500L, 1000L and 2000L and also in our Mobius® MIX and Mobius® Power MIX Single-Use
assemblies and Mobius® iFlex Bioreactor assemblies.

Application

Buffer, media, product transportation, Intermediate fluid transfer and storage

Features and Benefits

Ultimus® Film technology provides enhanced bag strength, improved durability and leak resistance.

Packaging

Pack of 1 assembly

Components

Bag: Ultimus® Film, Polyolefin
Tubing: TPE, Platinum-cured sillicone.
Coupler: PP
Luer: PP
Adapter SterlEnz: PP, Silicone
Plug end: PP.
Pinch clamp: Polyester, PBT

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTIMUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Film selection is guided by process needs, compatibility of the process fluid with film, process volume and risk. This brief compares the properties of our films so you can make the best selection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service