Skip to Content
Merck

Skip To

TA-5ML-B

Blood typing BIOSCOT IVD Reagent RHCE (Rh) Specificities

BIOSCOT Anti-E from the clone MS-80/MS-258, Human IgM, Blood Grouping Reagent

Synonym(s):

Anti-E (clone MS-80/MS-258), Blood Grouping Reagent

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
41116102
Clone:
MS-80/MS-258, monoclonal
Technique(s):
agglutination assay: suitable
Application:
Agglut.
Citations:
-
Note: For further information on price, availability or to place an order, please contact us


biological source

human

Quality Segment

clone

MS-80/MS-258, monoclonal

packaging

pkg of 5 mL

manufacturer/tradename

BIOSCOT

storage condition

do not freeze

concentration

18-40 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.1-7.3

isotype

IgM

application(s)

clinical testing

shipped in

ambient

storage temp.

2-8°C

General description

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 36 months from date of filtration

CE marked under the requirements of EU regulation 2017/746

For In Vitro use only .

Application

For use by Tube;Slide;Microplate;Gel and Column techniques
Vialled, Labeled

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
TA-10X5ML-UTA-10X5ML-BTA-1L-BK
clone

MS-80/MS-258, monoclonal

clone

MS-80/MS-258, monoclonal

clone

MS-80/MS-258, monoclonal

clone

MS-80/MS-258, monoclonal

biological source

human

biological source

human

biological source

human

biological source

human

isotype

IgM

isotype

IgM

isotype

IgM

isotype

IgM

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

600

Quality Level

600

Quality Level

600

Quality Level

600

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library