Skip to Content
Merck
All Photos(1)

Key Documents

SYMBCHH01

Millipore

Steritest® Symbio communication hub holder

For hoods, for use with Steritest® Symbio pump

Synonym(s):

Mount

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
CZK 23,500.00

CZK 23,500.00


In StockDetails


Request more information

Select a Size

Change View
1 UNIT
CZK 23,500.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

CZK 23,500.00


In StockDetails


Request more information

material

stainless steel

Agency

EP (2.6.1)
JP (4.06)
USP (71)

packaging

pkg of 1 unit

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® Symbio communication hub holder for hoods is an optional accessory for Steritest® Symbio Pump. The communication hub is not intended to be placed in the laminar flow hood, and hence can be fixed to one of the laminar flow hood feet using the communication hub holder for hoods. This accessory frees the space on the floor and its unique design is compatible with many types of support rods.

Application

Steritest® Symbio communication hub holder for hoods is used to attach the communication hub to one of the legs of the laminar flow hood. The depth can be adjusted.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service