Skip to Content
Merck
All Photos(1)

Key Documents

SX0650

Supelco

Sodium Molybdate, Dihydrate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Sodium Molybdate, Dihydrate, Disodium Molybdate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MoNa2O4 · 2H2O
CAS Number:
Molecular Weight:
241.95
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-551-7
grade:
ACS reagent
form:
powder
solubility:
840 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

meets USP/NF

Assay

99.5-103.0%

form

powder

color

white

pH

9-10 (20 °C, 840 g/L in H2O)

mp

687 °C

solubility

840 g/L

anion traces

chloride (Cl-): ≤0.005%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.015%

cation traces

Fe: ≤0.001%
NH4+: ≤0.001%
heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

room temp

InChI

1S/Mo.2Na.4O/q;2*+1;;;2*-1

InChI key

TVXXNOYZHKPKGW-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Ammonium (NH4)0.001% maxAssay99.5-103.0%Chloride (Cl)0.005% maxColorWhiteFormPowderHeavy metals (as Pb)5ppm maxInsoluble matter0.005% maxIron (Fe)0.001% maxPhosphate (PO4)5ppm maxSulfate (SO4)0.015% maxpH of a 5% solution at 25°C7.0-10.5

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service