Skip to Content
Merck
All Photos(1)

Key Documents

SX0205

Supelco

Silver nitrate

crystals, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Lunar Caustic, Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-853-9
form:
solid
solubility:
2160 g/L
Pricing and availability is not currently available.

vapor density

5.8 (vs air)

Quality Level

Assay

≥99.0%

form

solid

impurities

≤0.01% Substances not precipitated by HCl

color

colorless

pH

5.4-6.4 (20 °C, 100 g/L in H2O)

bp

444 °C/1013 hPa (decomposition)

mp

212 °C (dec.) (lit.)

solubility

2160 g/L

anion traces

chloride (Cl-): ≤5 ppm
sulfate (SO42-): ≤0.002%

cation traces

Cu: ≤2 ppm
Fe: ≤2 ppm
Pb: ≤0.001%

shipped in

ambient

storage temp.

room temp

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay: 99.0% min
Chloride (Cl): 5ppm max
Clarity of solution: To pass test
Color: Colorless
Copper (Cu): 2ppm max
Form: Crystals
Free acid: To pass test
Iron (Fe): 2ppm max
Lead (Pb): 0.001% max
Substances not precipitated by HCl: 0.01% max
Sulfate (SO4): 0.002% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service