Skip to Content
Merck
All Photos(1)

Key Documents

ST1553

Sigma-Aldrich

Anti-VCP Mouse mAb (4A8)

liquid, clone 4A8, Calbiochem®

Synonym(s):

Anti-Valosin-Containing Protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 11,700.00

CZK 11,700.00


Estimated to ship onMay 05, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 11,700.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 11,700.00


Estimated to ship onMay 05, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A8, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VCP(7415)

General description

Purified mouse monoclonal antibody. Recognizes the ~67 kDa VCP protein.
Recognizes the ~67 kDa VCP protein in HePG2 cells and human placenta tissue.
This Anti-VCP Mouse mAb (4A8) is validated for use in ELISA, Immunoblotting, Paraffin Sections for the detection of VCP.

Immunogen

Full-length, recombinant human VCP (aa 1-589), expressed as a GST fusion protein

Application



ELISA (see comments)
Immunoblotting (1-5 g/ml)
Paraffin Sections (3 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
HePG2 cells, human placenta tissue

Other Notes

For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp Voisard et al.
International journal of molecular sciences, 23(12) (2022-06-25)
Valosin-containing protein (VCP) acts as a key regulator of cellular protein homeostasis by coordinating protein turnover and quality control. Mutations in VCP lead to (cardio-)myopathy and neurodegenerative diseases such as inclusion body myopathy with Paget's disease of the bone and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service