Skip to Content
Merck

Skip To

ST1504

Anti-TCTP Rabbit pAb

liquid, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 μL

Estimated to ship on09 September 2026fromAreál Kühne+Nagel spol. s r.o.

9 120,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
human
Application:
Citations:
-

9 120,00 CZK


Estimated to ship on09 September 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Immunoblotting (1:1000))

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPT1(7178)

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the ~23 kDa TCTP protein.
Recognizes ~23 kDa TCTP protein in U937 cells.
This Anti-TCTP Rabbit pAb is validated for use in Immunoblotting for the detection of TCTP.

Immunogen

recombinant, human TCTP expressed in E. coli

Application

Immunoblotting (1:1000)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Regulatory Review (Z)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DR1071CB1047DR1103
description

liquid, Calbiochem®

description

liquid, Calbiochem®

description

liquid, Calbiochem®

description

liquid, Calbiochem®

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

serum

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

liquid

form

liquid

form

liquid

form

liquid

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters