SSCSHB3A01
Lynx® S2S
3/8" Lynx® S2S Connector, 3/8" hose barb inlet and 3/8" hose barb outlet
Synonym(s):
Sterile to sterile single-use, single actuation connector
Select a Size
CZK 3,230.00
Estimated to ship onFebruary 28, 2025Details
Select a Size
About This Item
CZK 3,230.00
Estimated to ship onFebruary 28, 2025Details
Recommended Products
material
polysulfone (21 CFR 177.1655)
silicone O-ring (21 CFR 177.2600)
silicone gasket (21 CFR 177.2600)
Quality Level
sterility
non-sterile
product line
Lynx®
parameter
27.3 L/min flow rate at 1.38 bar (20 psi) and 25 °C
4-40 °C operating temperature (39-104 °F)
4.1 bar max. pressure (60 psi) (operating pressure)
fitting
3/8 in. inlet hose barb for 0.25 in (7.1 mm) I.D.
3/8 in. outlet hose barb for 0.25 in (7.1 mm) I.D.
General description
The SSCSHB3A01 Lynx S2S connector set is constructed of a female coupling and a male coupling. With these two components assembled, the sterile fluid path is enabled. Once assembled, the two components cannot be disassembled, reducing the risk of sterility breach.
This Lynx S2S unit has a 3/8 inch. diameter and a hose barb inlet/outlet connection.
Safe, Disposable Connectors
The Lynx S2S connector is disposable and intended for single-use, one-time actuation. There are no parts to clean-in-place, and the risk of contamination is reduced.
Sterility Assurance During Actuation
The Lynx S2S connector is 100% air-integrity tested in manufacturing. Connector sets were tested utilizing an aerosolized bacterial challenge method with 106 B. diminuta and direct bacterial soiling method with B. diminuta, assuring a sterile connection can be made in non-classified areas.
Application
Preparation Note
Gamma compatible up to 45 kGy; Autoclave once for 30 minutes @ 130 °C. Do not steam in place.
Legal Information
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service