Skip to Content
Merck
All Photos(1)

Documents

SIPK0SIX2

Milli-Q

SimpliPak® Purification Cartridge

For Simplicity® systems with DI water feed

Synonym(s):

SimpliPak® 2 Purification Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

material

resin (Electronic Grade Ion-Exchange Resin)

Quality Level

feature

can remove trace contaminants

packaging

pkg of 1 unit

manufacturer/tradename

SimpliPak®

resistivity

>18.0 MΩ-cm (at 25 °C)

compatibility

for use with Simplicity® UV
for use with Simplicity®

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

SimpliPak® 2 Purification Cartridge is designed to remove ionic and organic contaminates down to trace levels from DI water feed. The cartridge is constructed with clean and high-purity materials to minimize the leaching of extractable contaminants.

Application

SimpliPak® 2 Purification Cartridge can be used with Simplicity® water purification systems.

Features and Benefits

  • Genuine consumable for the replacement of SIPK0SIX2 - SimpliPak® 2 Purification Cartridge.
  • Care-free maintenance: easily installed and replaced.
  • Generates ionic and organic contaminant-free water by ion exchange and activated carbon technologies

Legal Information

SIMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
SIMPLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service