Skip to Content
Merck

SIMPAK0D2

SimPak® Purification Cartridge

For use with Simplicity® and Synergy® water purification systems with DI water feed

Synonym(s):

SimPak® 2 Purification Cartridge

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
33050190
UNSPSC Code:
41104212
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 unit

manufacturer/tradename

SimPak®

compatibility

for use with Simplicity® (1999-2006), for use with Synergy® (2001-2006)

shipped in

ambient

General description

SimPak® 2 Purification Cartridge is designed to remove ionic and organic contaminates down to trace levels from DI water feed. The cartridge is constructed with clean and high-purity materials to minimize the leaching of extractable contaminants.

Application

SimPak® 2 Purification Cartridge can be used with Simplicity® and Synergy® water purification systems.

Features and Benefits

  • Genuine consumable for the replacement of SIMPAK0D2 - SimPak® 2 Purification Cartridge.
  • Generates ionic and organic contaminant-free water by ion exchange, activated carbon, and depth filtration technologies

Legal Information

SIMPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
SIMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
SYNERGY is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under SimPak® Purification Cartridge




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library