Skip to Content
Merck

Skip To

SCT281

BioTracker Blue Live Cell Cycle Assay Solution

Human

Synonym(s):

cell cycle dye, cell cycle indicator, cell cycle probe, live cell probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 vial

Estimated to ship onJune 04, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 3,520.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.47
Biological source:
human
Manufacturer/tradename:
Millipore

CZK 3,520.00


Estimated to ship onJune 04, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

BioTracker Blue Live Cell Cycle Assay Solution,

biological source

human

Quality Segment

packaging

vial of 50 tests

manufacturer/tradename

Millipore

technique(s)

cell culture | stem cell: suitable, flow cytometry: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Propidium iodide (PI) is often used for cell cycle analysis by flow cytometry. Compared to this analysis method, the BioTracker Far Red Live Cell Cycle Assay Solution does not require cell fixation or permeabilization, and is permeable in live cells. Use 5 uL of probe solution in 500 uL for each sample. Vial contains enough for 50 tests.IMPORTANT APPLICATION NOTES:*Wash cells thoroughly to remove trypsin. Residual trypsin can confound analysis of flow cytometry populations.*Adjust gates after cells are stained with probe SPECTRAL PROPERTIESExcitation: 405 - 407 nmEmission: 450/50 BPUse violet laser for excitation.
The control mechanism of the cell cycle is closely linked to cell proliferation. The cell cycle includes cell division, in which one cell produces two daughter cells. The cell cycle can be assessed by detecting cells stained for DNA using flow cytometry, followed by analysis of the proportion of stained nuclei in cell cycle phase. The cell cycle is classified into two major phases: interphase and the mitotic (M) phase. Interphase is composed of G1 phase (preparation for DNA synthesis which occurs prior to cell division), S phase (DNA synthesis and replication), and G2 phase (chromosome duplication). M phase involves mitosis and cytokinesis. Cell cycle progression is regulated by cyclins and cyclin-dependent kinases. To determine the action of drugs such as anticancer therapeutics, it is essential to analyze the mechanism of cell-cycle checkpoint.

Features and Benefits

Blue fluorescence probe for analysis of the cell cycle by flow cytometry.

Preparation Note

Store BioTracker Blue Live Cell Cycle Assay Solution at -20 °C, protected from light.

Other Notes

Live cell probe for fluorescent cell cycle analysis. Unlike propidium iodide (PI), this probe is membrane permeable and highly specific for DNA, which makes it possible to analyze the cell cycle by simply adding the reagent to a cell suspension without any fixation, permeabilization, or other sample prep.

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


pictograms

CorrosionEnvironment

signalword

Danger

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Aquatic Chronic 2 - ED ENV 1 - Eye Dam. 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9016-45-9

CAS No.

9016-45-9

CAS No.

9016-45-9

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters