Skip to Content
Merck
All Photos(2)

Documents

SCT248

Sigma-Aldrich

BioTracker Putrescine Polyamine Green Fluorescent Live Cell Probe

Human

Synonym(s):

BioTracker Clickable Polyamine Putrescine Live Cell Probe, clickable polyamine probe, clickable putrescine probe, live cell probe, polyamine probe, polyamine transport system probe, putrescine probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

product name

BioTracker Putrescine Polyamine Green Fluorescent Live Cell Probe,

biological source

human

Quality Level

packaging

vial of 500 μg

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

The relationship between polyamine systems and cancer---plus critical normal cell processes--- led to the creation of clickable polyamine derivatives which are innovative and novel tools for functional studies of the polyamine transport system (PTS). Putrescine and other important polyamine derivatives were conjugated to a BODIPY fluorophore through click chemistry.MW: 558.34 g/mol SPECTRAL PROPERTIESλex: 494 nmλem: 508 nm

Features and Benefits

The BioTracker Clickable Polyamine Putrescine Live Cell Probe is click-conjugated to a green fluorophore to enable the study of polyamine transport with superior cellular uptake.Probe may be dissolved in DMSO to make a 10mM stock solution. 1 uL of a 10 mM stock solution may be added to 1 mL media to produce a 10 uM working solution. Add probe/media solution to washed cells and incubate at 37C for up to 4h.

Target description

Polyamines (putrescine, spermine, spermidine) play a variety of critical roles in the cell. Some of these processes include gene expression, cell proliferation and differentiation, and cellular stress management. Polyamine dysregulation is key in a variety of cancers. In colorectal cancer, polyamine content and biosynthesis is significantly higher when compared to normal colorectal tissue. A relationship between polyamines and their metabolites has been used to observe progression in cancers of the breast, lung, colorectal, ovarian, prostate, and pancreas. In normal cells, there is tight regulation of polyamines through normal cellular mechanisms. One of these is the polyamine transport system (PTS). The characteristics of the PTS are not fully understood, but study of the PTS has shown that many cancer cells have high PTS activity, suggesting this system as a promising target for cancer research.

Physical form

red-brown solid

Storage and Stability

Store at -20 °C, protected from light

Other Notes

This putrescine derivative click-conjugated to BODIPY is optiized for the study of the polyamine transport system, which is exploited in diverse cancers.

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roeland Vanhoutte et al.
Chembiochem : a European journal of chemical biology, 19(9), 907-911 (2018-02-17)
Polyamines are essential for cell growth and differentiation, but their trafficking by the polyamine transport system is not fully understood. Herein, the synthesis of several azido-derivatized polyamines for easy conjugation by click chemistry is described. Attachment of a 4,4-difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY)
Norin Nabil Hamouda et al.
The Journal of biological chemistry, 296, 100182-100182 (2020-12-15)
Polyamines, such as putrescine, spermidine, and spermine, are physiologically important polycations, but the transporters responsible for their uptake in mammalian cells remain poorly characterized. Here, we reveal a new component of the mammalian polyamine transport system using CHO-MG cells, a
Sarah van Veen et al.
Nature, 578(7795), 419-424 (2020-01-31)
ATP13A2 (PARK9) is a late endolysosomal transporter that is genetically implicated in a spectrum of neurodegenerative disorders, including Kufor-Rakeb syndrome-a parkinsonism with dementia1-and early-onset Parkinson's disease2. ATP13A2 offers protection against genetic and environmental risk factors of Parkinson's disease, whereas loss

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service