Skip to Content
Merck

Skip To

SCT241

BioTracker LD-1 Near-IR Lipid Droplet Live Cell Probe

Synonym(s):

Lipid Droplet Live Cell Dye, Lipid Droplet Live Cell Probe, Lipid Droplet Probe, NIR Lipid Droplet Dye

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 vial
Check Cart for Availability
CZK 5,770.00

About This Item

NACRES:
NA.31
UNSPSC Code:
41106514
Manufacturer/tradename:
Millipore

CZK 5,770.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

vial of 1 mg

Quality Segment

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable, flow cytometry: suitable

detection method

fluorometric

shipped in

dry ice

storage temp.

−20°C

General description

Lipid droplets are fat storage organelles that play a key role in diverse cell functions. These include energy homeostasis, cellular metabolism, and innate immunity (1). As research in cellular lipid storage has progressed, it has become apparent that lipid droplet abnormalities may serve as markers for dysregulated cell physiology. This observation has come to be utilized for cancer diagnosis. Fluorescent probes continue to be promising tools for imaging due to their selectivity, sensitivity, and capabilities for real-time testing (1). Lipid droplet probes developed to date may suffer from suboptimal selectivity as well as “self-absorption.” The LD-1 lipid droplet probe was developed with a near-infrared (NIR) fluorophore to provide enhanced tissue penetration and to minimize autofluorescence. The probe fluorophore was designed to turn on through intramolecular charge transfer (ICT) and motion-induced changes in emission (MICE) (1). The fluorophore spectral characteristics include a large Stokes shift which helps to reduce probe self-absorption. The unique probe structure enables increased emission output in a high-viscosity environment.
Recent years have seen increased demand for exceptionally sensitive and specific fluorescent probes for imaging lipid droplets (LD)s, particularly in the context of cancer diagnosis.

This LD-1 probe is chemically optimized for the low-polarity, high-viscosity microenvironment found in lipid droplets.

In contrast with Raman microscopy and transmission electron microscopy (TEM) fluorescent small molecule probes have been developed as promising tools for detecting LDs . Advantages of fluorescent LD probes include the capability for real-time and in vitro testing, plus excellent selectivity and sensitivity.


Spectral properties

Absorbance : 650 nm
Emission (max): 745 nm

Reference

Wu X, Wang X, Li Y, Kong F, Xu K, Li L, Tang B. 2022. A near-infrared probe for specific imaging of lipid droplets in living cells. Anal Chem. 94(11):4881–4888.

Application

Purity > 98% confirmed by HPLC. Identification confirmed by HNMR, LC-MS, and elemental analysis.

Features and Benefits

This near-infrared (NIR) fluorescent probe selectively binds lipid droplets in live cells using a rapid, simple protocol for fluorescence microscopy or flow cytometry detection.

Physical form

Lyophilized. Black solid.

Preparation Note

Store BioTracker LD-1 Near-IR Lipid Droplet Live Cell Probe at -20°C, desiccated and protected from light. Centrifuge vial briefly to collect contents at bottom of vial before opening.

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCT089SCT066SCT144
technique(s)

cell culture | mammalian: suitable, flow cytometry: suitable

technique(s)

-

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

shipped in

dry ice

shipped in

ambient

shipped in

-

shipped in

ambient

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

manufacturer/tradename

Millipore

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

Quality Level

100

Quality Level

200

Quality Level

-

Quality Level

-

detection method

fluorometric

detection method

-

detection method

-

detection method

fluorometric


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters