Skip to Content
Merck

SCT111

BioTracker 405 Blue SE Cell Proliferation Kit

Live cell imaging cell proliferation kit used for fluorescent cell labeling in flow cytometry applications.

Synonym(s):

Live cell imaging probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

storage temp.

-10 to -25°C

General description

BioTracker SE Cell Proliferation Kits diffuse passively into live cells and are used for long-term cell labeling. They are initially non-fluorescent esters of amine-reactive dyes, but are converted to fluorescent dyes by intracellular esterases. The dyes then covalently react with amine groups on intracellular proteins, forming fluorescent conjugates that are retained in the cell. Immediately after staining a single, bright fluorescent population will be detected by flow cytometry. Each cell division that occurs after labeling is revealed by the appearance of a successively dimmer fluorescent peak on a flow cytometry histogram (Fig. 1). Cell proliferation dyes can be used to track cell divisions in vivo or in vitro. The staining can withstand fixation and permeabilization for subsequent immunostaining. Alternative applications of cell proliferation dyes include uniform labeling of cell cytoplasm for microscopy, or labeling cells for quantitation of cell number by microplate reader.

Spectral Properties
Absorbance: 408nm
Emission: 452nm

Application

Live cell fluorescent imaging
Live cell imaging cell proliferation kit used for fluorescent cell labeling in flow cytometry applications.
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Physical form

Liquid

Preparation Note

Store BioTracker 405 Blue SE Cell Proliferation Kit -20ºC. Protect From Light.
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening. Ideally the 5 mM DMSO stock solution should be prepared on the day of use. Aliquots may be stored for later use, but activity may be reduced over time. The dyes should only be added to aqueous buffer immediately before staining.

Analysis Note

Spectral Properties
Absorbance: 408nm
Emission: 452nm

Other Notes

1. 10 vials of BioTracker 405 Blue SE Dye (100nmol) (CS224590)

2. 1 vials of Anhydrous DMSO (500µL) (CS224578)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library