Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.21
UNSPSC Code:
12171501
type
for cell culture
technique(s)
cell based assay: suitable
detection method
fluorometric
General description
442.39 g/mol
Application
Live cell fluorescent imaging
Physical form
Lyophilized
Preparation Note
Store BioTracker™ Blue Hydrogen Sulphide Live Cell Dye at -20°C, desiccate and protect from light Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.
Analysis Note
Purity: ≥ 92% confirmed by HNMR, LC-MS and HPLC and elemental analysis
Molar Mass: 493.43 g/mol
Molar Mass: 493.43 g/mol
Legal Information
BioTracker is a trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


