Skip to Content
Merck

Skip To

SCT070

BioTracker NIM-7 Lipids and Lysosomes Live Cell Dye

Live cell imaging fluorescent probe for simultaneous dual-color tracking of lipid droplets and lysosomes

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized

application(s)

cell analysis

detection method

fluorometric

General description

Lipid droplets (LDs) and lysosomes are specialized subunits within cells. LDs are the most hydrophobic sub-structures within cells and serve to store lipids for energy generation and membrane synthesis. Lysosomes are typically acidic organelles (pH 4.5–5.0) that play a critical role in the digestion and clearance of endocytosed and endogenous intracellular materials.BioTracker NIM-7 Lipids and Lysosomes Live Cell Dye is a naphthalimide-based fluorescent probe that allows lipid droplets and lysosomes to be labelled simultaneously and visualized separately using different excitation and detection channels. Lipid droplets and lysosomes can be visualized in the yellow and red fluorescence regions, respectively. The dye was found to have low cytotoxicity in multiple cell lines after 24 hour incubation. The dye also high photostability and can maintain 90% of original fluorescence intensity after 15 min of irradiation.Spectral properties:Lipid droplets: Excitation: 488 nm. Emission: 530-570 nm.Lysosomes: Excitation: 561 nm. Emission: 640-680 nm.Research Category:Live Cell DyeLipid Signaling

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters