Skip to Content
Merck
All Photos(1)

Key Documents

SCMK001-S

Sigma-Aldrich

EPIGRO® Human Epidermal Keratinocyte Supplement Kit

Sign Into View Organizational & Contract Pricing

Select a Size

SCMK001-S
CZK 3,290.00

CZK 3,290.00


Estimated to ship onApril 16, 2025Details


Request a Bulk Order

Select a Size

Change View
SCMK001-S
CZK 3,290.00

About This Item

UNSPSC Code:
12352200

CZK 3,290.00


Estimated to ship onApril 16, 2025Details


Request a Bulk Order

Quality Level

Legal Information

EPIGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service