Skip to Content
Merck

Skip To

SCM142

PLTMax Human Platelet Lysate

PLTMax Human Platelet Lysate is a growth factor rich supplement that is a superior alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) culture. 500ml.

Synonym(s):

PLTMax hPL, hPL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 48,100.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.71
eCl@ss:
32011203

CZK 48,100.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type mesenchymal stem cell(s)

shipped in

ambient

storage temp.

-65 to -96°C

General description

PLTMax Human Platelet Lysate is a growth factor rich supplement that is a superior alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) culture. Optimal growth of MSCs can be achieved with 5% PLTMax Human Platelet Lysate. PLTMax Human Platelet Lysate has been used to effectively to grow mesenchymal stem cells derived from other species including mouse, rabbit and porcine.



PLTMax Human Platelet Lysate is derived from normal human donor platelets collected at U.S. blood centers. Multiple donors units are pooled in large batch sizes and manufactured to produce a consistent product. Heparin (porcine) approved for human intravenous use is used in the preparation of PLTMax Human Platelet Lysate. This product contains human derived materials and must be treated as potentially infectious. Universal precautions for biological samples should be used in handling and disposal.

Application

Instructions for Use
• Add Heparin to the media at a final concentration of 2 U/mL. Failure to add heparin may result in coagulation of certain media.
• For optimal growth of human mesenchymal stem cells, add 5% vol/vol PLTMax Human Platelet Lysate to usual cell culture medium such as DMEM and αMEM.
• Plate cells at approximately 1.5x10^3 cells per cm2.
• Do not allow primary MSC confluence to exceed 80%.

Particulate Formation
• Product has been aseptically filtered with 0.2 µm filters. Particulates will be observed after thawing. Particulate formation is normal will not affect performance even in the presence of large particulates. Filtration is not recommended.
• To reduce particulates from forming, minimize freeze/thaw cycles and extended storage at 4°C. Some particulate formation can be reversed with brief incubation at 37°C, if desired.

Analysis Note

PLTMax Human Platelet Lysate is manufactured with the highest quality of raw materials. All human derived materials are obtained from government inspected facilities and are of US origin. Each donor unit was approved for human use and has been tested for infectous diseases including HIV-1, HIV-2, HIV-NAT, HCV, HBsAg, and RPR for Syphilis.

Each lot is extensively tested for the following parameters:
pH: 6.8-7.8
Total protein: 4.0-6.5 g/dl
Endotoxin USP <8>: <10 EU/mL
Mycoplasma: Not detected
Sterility Testing: Negative for growth

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H9661P4306P3201
form

liquid

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

biological source

human

biological source

human

biological source

human

biological source

human

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-

storage temp.

-65 to -96°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under PLTMax Human Platelet Lysate


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. PLTMaxを使用して実験をしているのですが、PRPとの性質の違いはありますか。一般的に使用されているPRPとの違いをご教示ください。(製法や、ヘパリン以外の添加物などあれば教えていただきたいです。)

    1 answer
    1. The main difference is that PRP contains "whole" platelets (a type of blood cell), while PLTMax is a platelet "Lysate" i.e., it is a platelet "extract" that contains platelet growth factors rather than whole platelet cells.

      The manufacturing process for SCM142 is summarized below. An appropriate lysis buffer is used to lyse the platelets to prepare the lysate/extract. Further details are proprietary.

      Please see the details below:

      1. PRP stands for Platelet-rich plasma.
      "Platelet-rich plasma consists of two elements: plasma, or the liquid portion of blood, and platelets, a type of blood cell that plays an important role in healing throughout the body. Platelets are well-known for their clotting abilities, but they also contain growth factors that can trigger cell reproduction and stimulate tissue regeneration or healing in the treated area. Platelet-rich plasma is simply blood that contains more platelets than normal.

      To create platelet-rich plasma, clinicians take a blood sample from the patient and place it into a device called a centrifuge that rapidly spins the sample, separating the other components of the blood from the platelets and concentrating them within the plasma".

      This information can be accessed in the link below:

      https://www.hopkinsmedicine.org/health/treatment-tests-and-therapies/plateletrich-plasma-prp-treatment#:~:text=Platelet%2Drich%20plasma%20(PRP)%20injections%20are%20gaining%20popularity%20for,healing%20in%20a%20specific%20area.

      2. The following information below is found for Product SCM142 on the Product Detail Page:

      PLTMax® Human Platelet Lysate is a growth factor-rich supplement that is a superior alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) culture. Optimal growth of MSCs can be achieved with 5% PLTMax Human Platelet Lysate. PLTMax Human Platelet Lysate has been used to effectively grow mesenchymal stem cells derived from other species, including mice, rabbits, and porcine.

      PLTMax Human Platelet Lysate is derived from normal human donor platelets collected at U.S. blood centers. Multiple donor units are pooled in large batch sizes and manufactured to produce a consistent product. Heparin (porcine), approved for human intravenous use, is used to prepare PLTMax Human Platelet Lysate. This product contains human-derived materials and must be treated as potentially infectious. Universal precautions for biological samples should be used in handling and disposal.

      Helpful?

Reviews

No rating value

Active Filters