Skip to Content
Merck

Skip To

SCM111

Human ES/iPS Neuronal Differentiation Medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 9,860.00

About This Item

UNSPSC Code:
41106200
NACRES:
NA.32
eCl@ss:
32160801

CZK 9,860.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

technique(s)

cell differentiation: suitable

General description

EMD Millipore′s Human ES/iPS Neuronal Differentiation Medium is a serum free/chemically defined optimized media for the rapid production of terminally differentiated neurons from human neural progenitor cells. The media relies on both small molecules neural inducers and supplements to generate a highly enriched population of terimally differentiated TUJ-1/MAP2ab positive end stage neurons based on establish protocols (1,2).

Preparation Note

1) Neuronal Differentiation Basal Medium (Part No. CS211007). One 100 mL bottle. 2) Neural Supplement 1 (50X) (Part No. CS210992). One 2.5 mL bottle. 3) Adenosine 3′, 5′-cyclic Monophosphate, N6, O2′-Dibutyryl-, Sodium Salt (Cat. No. 28745-25mg). One vial containing 25 mg of lyophilized powder. 4) Ascorbic Acid 2-Phosphate (Part No. 2004011). One 0.5 mL vial of 100 mM solution.
Store at -20C. Good for 3 months from date of receipt or until expiration date on bottle when reagents are handled and stored appropriately. Please note: The supplemented ES/iPS Neuronal Differentiation Medium should be stored at 2 to 8C for up to 1 week.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library