Skip to Content
Merck
All Photos(1)

Key Documents

SCM037

Sigma-Aldrich

FibroGRO® Xeno-Free Human Fibroblast Expansion Medium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CZK 3,860.00

CZK 3,860.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 ML
CZK 3,860.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

CZK 3,860.00


In StockDetails


Request a Bulk Order

form

liquid

Quality Level

manufacturer/tradename

Chemicon®
Millipore

technique(s)

cell culture | mammalian: suitable

input

sample type epithelial cells

shipped in

dry ice

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
FIBROGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service