Skip to Content
Merck
All Photos(2)

Key Documents

SCC352

Sigma-Aldrich

3dGRO Duodenum Intestinal Organoids, Age 50 (Prep ht-188-D)

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
CZK 57,700.00

CZK 57,700.00


Estimated to ship onMarch 14, 2025Details



Select a Size

Change View
1 VIAL
CZK 57,700.00

About This Item

UNSPSC Code:
41106509
NACRES:
NA.21

CZK 57,700.00


Estimated to ship onMarch 14, 2025Details


packaging

vial of >1500 viable organoids cells

Quality Level

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell culture | stem cell: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

Application

  • Viability: >1500 viable organoids/vial
  • Organoid Growth: Pass
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Mycoplasma Contamination: Negative
  • STR Profile: Pass

Patient derived organoids (PDOs) are novel in vitro 3D cell models that preserve original tissue physiology and molecular pathology, thus representing a clinically-relevant alternative to traditional 2D cell lines and an effective tool to refine and reduce animal models. PDOs can be derived from adult patient biopsies or resected tissues containing native LGR5+ stem cell populations and cultured within an ECM-rich substrate using specialized organoid media such as L-WRN conditioned media. Adult tissue derived organoids are phenotypically and genetically stable in long term culture, presenting more mature phenotypes compared to iPSC-derived organoids. Importantly, PDOs have shown to be able to predict patient clinical responses to chemotherapeutics.We are now offering a comprehensive biobank of highly characterized tissue-derived human gastrointestinal organoids from normal and diseased patients. The intestinal organoid biobank contains over 50 highly characterized intestinal organoids from both normal and diseased patients derived from multiple regions of the digestive systems including small intestine (duodenum, ileum), stomach, rectum and colon.Cell Line Characteristics• Sex: Female• Age: 50 Years • Organ: Duodenum• Disease: Normal

Features and Benefits

Cryopreserved patient derived gastrointestinal organoids (PDOs)

Storage and Stability

Store in liquid nitrogen. The organoids can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

3dGRO is a trademark of Merck KGaA, Darmstadt, Germany.
3dGRO organoids were derived utilizing HUB Organoid Technology.
The purchaser of this product shall agree to HUB′s terms of use, which shall be separately acknowledged and accepted by such purchaser, prior to transfer of this product to purchaser.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service