Skip to Content
Merck

SCC153

9HTEo- Human Tracheal Epithelial Cell Line

Synonym(s):

9/HTEo, 9/HTEo-, 9HTE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32011203
UNSPSC Code:
41106514
NACRES:
NA.81
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

General description

9HTEo- is a human tracheal epithelial cell line that was immortalized with the origin-of-replication defective SV40 plasmid (pSVori-). The cell line retains characteristic features of normal differentiated tracheal epithelial cells including a cobblestone morphology, cytokeratin expression and the ability to form tight junctions.

9HTEo- cells contain the wild-type cystic fibrosis transmembrane conductance regulator (WT-CFTR) and exhibits functional cAMP-dependent chloride ion transport and calcium ion-dependent chloride transport (1,2).

9HTEo- is a useful model for the study of epithelial ion transport, secretion and biochemistry and frequently used as a wild-type control for human CF tracheobronchial epithelial cells such as CFBE41o- (Cat. No. SCC151) and CFTE29o- (Cat. No. SCC162).

Application

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact [email protected].

Analysis Note

Each vial contains more than 1X106 viable cells.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
SCC15304054839480447