Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
eCl@ss:
32160702
UNSPSC Code:
12352203
antibody form
affinity purified immunoglobulin
Quality Segment
Storage Class
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.