Skip to Content
Merck
All Photos(1)

Key Documents

S7908

Sigma-Aldrich

Amplifluor® SNPs HT Genotyping System FAM-SR

Sign Into View Organizational & Contract Pricing

Select a Size

5000 ASSAYS
CZK 91,500.00

CZK 91,500.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

Select a Size

Change View
5000 ASSAYS
CZK 91,500.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.84

CZK 91,500.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

application(s)

genomic analysis

Quality Level

detection method

fluorometric

General description

The Amplifluor SNPs HT Genotyping System FAM-SR is designed for users familiar with Amplifluor and is for use with fluorescent plate readers.

Legal Information

Amplifluor is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service