Skip to Content
Merck
All Photos(2)

Key Documents

S2HVU02RE

Millipore

Millipore® Stericup® Quick Release Vacuum Filtration System

pore size 0.45 μm, PVDF membrane, funnel capacity 250 mL, pack of 12 ea

Synonym(s):

SCHVU02RE, Stericup Quick Release-HV Sterile Vacuum Filtration System, sterile vacuum filter unit, sterile vacuum filtering unit, vacuum filtration system

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
CZK 3,450.00

CZK 3,450.00


Estimated to ship onMay 27, 2025Details


Request more information

Select a Size

Change View
12 EA
CZK 3,450.00

About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24

CZK 3,450.00


Estimated to ship onMay 27, 2025Details


Request more information

material

PVDF membrane
polystyrene bottle
polystyrene funnel(s)
polystyrene housing

Quality Level

sterility

sterile; irradiated

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

parameter

45 °C max. temp.

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

164 mm

capacity

250 mL

diam.

12.5 cm

filter diam.

73 mm

filtration area

40 cm2

funnel capacity

250 mL

matrix

Durapore®

pore size

0.45 μm pore size

fitting

inlet funnel
outlet flask (Receiver)

shipped in

ambient

General description

This item is a direct replacement for SCHVU02RE
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

Stericup® and Steritop® vacuum sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.

This Stericup® 0.45 μm pore size polyvinylidene fluoride (PVDF) membrane filter unit combines a 250 mL bottle-top with a 250 mL receiver flask and cap for processing and storage.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.

  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • The Quick Release filter funnel disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–10 of 15 Questions  
  1. What is the maximum processing volume for Stericup® and Steritop® filters? Can I add more volume after initial filtration?

    1 answer
    1. All Stericup® and Steritop® filters were developed for single-use application. The maximum volume is dependent on the receiver bottle capacity. Stericup® filters come with a receiver bottle with a specified maximum volume. For Steritop® filters, the maximum recommended volume is 2 L with an appropriate-sized receiver bottle. Larger processing volumes can result in membrane fouling, increase chance of spillage and contamination, and lead to possible airlock*.
      *Note that devices with PES membranes can airlock if filtered to dryness. Some liquid must be left on top of the filter, then the system stopped, receiver changed or emptied, and more sample volume added before restart. This increases the chance of spillage.

      Helpful?

  2. What is the chemical compatibility of the various Stericup® and Steritop® filters?

    What is the chemical compatibility of the various Stericup® and Steritop® filters?

    1 answer
    1. Helpful?

  3. Can Stericup® and Steritop® filters be used to prepare solutions that will be administered to patients?

    1 answer
    1. No, all Stericup® and Steritop® devices are Research Use Only.

      Helpful?

  4. Are Stericup® and Steritop® filters GMP-certified?

    1 answer
    1. No, Stericup® and Steritop® devices are not GMP-approved.

      Helpful?

  5. Which type of pump should be used with Stericup® and Steritop® vacuum filters?

    Which type of pump should be used with Stericup® and Steritop® vacuum filters?

    1 answer
    1. We recommend our Chemical Duty Pump: https://www.sigmaaldrich.com/product/mm/wp6111560

      Helpful?

  6. What Bacterial Endotoxins Test is used?

    1 answer
    1. The test method used complies with USP 85 and EP 2.6.14 to ensure the absence of endotoxin in our products.

      Helpful?

  7. Can Stericup® bottles be placed inside freezers for long-term storage of media?

    1 answer
    1. You can successfully freeze and store many aqueous solutions (such as culture media) in Stericup® bottles at temperatures up to -20 °C (-4 °F). It is strongly recommended that you run a sample stability trial under your actual storage conditions prior to using Stericup® bottles for frozen storage.

      Helpful?

  8. What tubing can be used with Stericup® and Steritop® vacuum filters?

    What tubing can be used with Stericup® and Steritop® vacuum filters?

    1 answer
    1. Stericup® and Steritop® filters are fitted with a 1/4” tube adapter. We recommend 3/16 in. (4.8 mm) ID silicone tubing to fit this tube adapter: https://www.sigmaaldrich.com/product/mm/xx7100004

      Helpful?

  9. What is the expiration date of Stericup® and Steritop® filters?

    1 answer
    1. Typically 3 years from sterilization.

      Helpful?

  10. What is the maximum vacuum pressure for filtration?

    1 answer
    1. 700 mm Hg (27.5” Hg, 0.9 bar) differential vacuum at 25 °C (77 °F)

      Helpful?

1–10 of 15 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service