Skip to Content
Merck
All Photos(1)

Key Documents

QIA113

Sigma-Aldrich

Caspase-8 Detection Kit (FITC-IETD-FMK)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CZK 19,900.00

CZK 19,900.00


Estimated to ship onMarch 11, 2026Details


Request a Bulk Order

Select a Size

Change View
1 KIT
CZK 19,900.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.84

CZK 19,900.00


Estimated to ship onMarch 11, 2026Details


Request a Bulk Order

usage

sufficient for 100 tests

Quality Level

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

technique(s)

flow cytometry: suitable

input

sample type intact cells

fluorescence

λex ~485 nm; λem ~535 nm

detection method

fluorometric

storage temp.

−20°C

General description

A fast, convenient, and sensitive method to measure caspase-8 in living cells. The fluorescent marker, FITC-IETD-FMK, is a labeled, cell-permeable, non-toxic inhibitor that binds irreversibly to activated caspase-8 in living cells. Caspase-8 can be detected by the measurement of fluorescence in a fluorescent microplate reader, flow cytometer, or by fluorescence microscopy.

Specificity

A broad range of species

Components

Labeled Caspase-8 Inhibitor (FITC-IETD-FMK), Wash Buffer, Unlabeled Caspase Inhibitor (Z-VAD-FMK), and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Specifications

Assay Time: 1.5 h

Storage and Stability

Upon arrival store the entire contents of the kit at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service