Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
H2Cl6Pt · 6H2O
Molecular Weight:
517.90
UNSPSC Code:
41116107
form
liquid
shipped in
ambient
storage temp.
no temp limit
General description
TITRISTAR
Analysis Note
Expiration date: Exp 24 months from mfg date
Assay: 10.0% min
Color: Yellow
Form: Clear liquid
Assay: 10.0% min
Color: Yellow
Form: Clear liquid
Storage Class
8B - Non-combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.