Skip to Content
Merck

Skip To

PLBC04310

Regenerated Cellulose Membrane Filter, 3 kDa NMWCO

Ultracel®, filter diam. 44.5 mm

Synonym(s):

Ultrafiltration Discs, 3 kDa NMW, Ultrafiltration membrane discs, Regenerated Cellulose Membrane Filter, 3 kDa NMWCO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 ea

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

7 610,00 CZK

About This Item

UNSPSC Code:
41104903
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
3 kDa NMWCO
Material:
regenerated cellulose membrane
Feature:
hydrophilic
Diam.:
44.5 mm
Sterility:
non-sterile

7 610,00 CZK


Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ultracel® 3 kDa Ultrafiltration Discs, Chemistry: Regenerated Cellulose (RC) Filter Diameter: 44.5 mm Amicon® Bioseparations

material

regenerated cellulose membrane

Quality Segment

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Ultracel®

technique(s)

protein clean-up: suitable (desalting), protein purification: suitable (concentration)

diam.

44.5 mm

filter diam.

44.5 mm

filtration area

13.4 cm2

matrix

Ultracel®

pore size

3 kDa NMWCO

shipped in

ambient

General description

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel regenerated cellulose and Biomax polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore PVDF cut discs are available 0.1 µm, 0.22 µm, and 0.45 µm pore sizes in both 63.5 mm and 75 mm diameters.

Features & Benefits:
•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
•Both Ultracel, Durapore, and Biomax membranes are available in a wide range of pore sizes to meet your separation requirements
•Compatible with Amicon Stirred Cells

Applications:
Concentration, Buffer Exchange, Desalting
Device Configuration: Ultrafiltration Discs
Filter Code: PLBC
Filter Type: Ultrafiltration

Application

3 kDa NMWL, Ultracel regenerated cellulose, 44.5 mm diameter

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PLBC02510PLBC06210PLBC09005
description

Chemistry: Regenerated Cellulose (RC) Filter Diameter: 44.5 mm Amicon® Bioseparations

description

Chemistry: Regenerated Cellulose (RC) Filter Diameter: 25 mm Amicon® Bioseparations

description

Chemistry: Regenerated Cellulose (RC) Filter Diameter: 63.5 mm Amicon® Bioseparations

description

Chemistry: Regenerated Cellulose (RC) Filter Diameter: 90 mm Amicon® Bioseparations

manufacturer/tradename

Ultracel®

manufacturer/tradename

Ultracel®

manufacturer/tradename

Ultracel®

manufacturer/tradename

Ultracel®

pore size

3 kDa NMWCO

pore size

3 kDa NMWCO

pore size

3 kDa NMWCO

pore size

3 kDa NMWCO

diam.

44.5 mm

diam.

25 mm

diam.

63.5 mm

diam.

90 mm

material

regenerated cellulose membrane

material

regenerated cellulose membrane

material

regenerated cellulose membrane

material

regenerated cellulose membrane

sterility

non-sterile

sterility

-

sterility

non-sterile

sterility

-

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters