Skip to Content
Merck
All Photos(1)

Key Documents

PK1010

Sigma-Aldrich

PhosphoDetect Anti-Syk (pTyr525/526) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
CZK 9,450.00

CZK 9,450.00


Estimated to ship onApril 28, 2025Details


Request a Bulk Order

Select a Size

Change View
50 μL
CZK 9,450.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 9,450.00


Estimated to ship onApril 28, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr525/pTyr526)

Gene Information

human ... SYK(6850)

General description

Protein A and immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~72 kDa Syk protein phosphorylated on Tyr525/526.
Recognizes the ~72 kDa Syk protein phosphorylated at Tyr525/526 in human Syk or Tyr519/520 in mouse Syk in 293T cells expressing wild-type Syk and CD8.
This PhosphoDetect Anti-Syk (pTyr525/526) Rabbit pAb is validated for use in ELISA, Immunoblotting for the detection of Syk (pTyr525/526).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr525/526 phosphorylation sites of human Syk

Application


ELISA (1:1000)
Immunoblotting (1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM sodium HEPES, 100 µg/ml BSA, 50% glycerol, pH 7.5.

Reconstitution

Do not aliquot.

Analysis Note

Positive Control
293T cells transfected with wild-type Syk and CD8

Other Notes

Coopman, P.J., et al. 2000. Nature406, 742.
Turner, M., et al. 2000. Immunol. Today21, 148.
Zang, J., et al. 2000. J. Biol. Chem.275, 35442.
Chu, D.H., et. al. 1998. Immunol. Rev.165, 167.
Cheng, A.M. and Chan, A.C. 1997. Curr. Opin. Immunol.9, 528.
Kurosaki, T. 1997. Curr. Opin. Immunol.9, 309.
Tyr525/526 represent the phosphorylation sites in human Syk, while Tyr519/520 are the equivalent phosphorylation sites in mouse Syk. For immunoblotting the membrane should be incubated with antibody diluted in 5% BSA, 1X TBS, 0.1% Tween®-20 detergent at 4°C overnight with gentle shaking. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyemin Lee et al.
Frontiers in pharmacology, 13, 763678-763678 (2022-02-22)
B lymphocyte-induced maturation protein-1 (Blimp-1) is a transcriptional repressor and plays a crucial role in the regulation of development and functions of various immune cells. Currently, there is limited understanding about the regulation of Blimp-1 expression and cellular functions in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service