Skip to Content
Merck

Skip To

PHCC0CLIP

Scepter O-ring Kit

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41104923
NACRES:
NB.22
eCl@ss:
32011202


manufacturer/tradename

Scepter

technique(s)

cell counting: suitable

shipped in

ambient

storage temp.

room temp

General description

After repeated use the Scepter O-ring may wear. It can be easily replaced with this kit and by following the instructions included in the manual.

Other Notes

2 O-rings

1 filter cover




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters