Skip to Content
Merck
All Photos(2)

Key Documents

PEZGS0496

Millipore

Millicell® EZ Slide

4-well glass chamber slide, 1.7 cm2/well, 96/pk, sterile

Synonym(s):

4 well glass slide, Chamber slides

Sign Into View Organizational & Contract Pricing

Select a Size

96 EA
CZK 22,600.00

CZK 22,600.00


Please contact Customer Service for Availability


Select a Size

Change View
96 EA
CZK 22,600.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22

CZK 22,600.00


Please contact Customer Service for Availability

material

polypropylene wells
polystyrene lid (and slide holder)
soda-lime glass slide

sterility

ethylene oxide treated
sterile

feature

Raised corner ridges on the lid facilitate stacking of multiple units

packaging

pack of 96 (6 boxes, 16 units/box)

manufacturer/tradename

Millicell®

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable
immunocytochemistry: suitable

L × W × H

75 mm × 25 mm × 1 mm, slide only
77.8 mm × 33.8 mm × 13.9 mm, excluding lid
77.8 mm × 33.8 mm × 16.2 mm, including lid

size

4 wells

well surface area

1.7 cm2

well volume

500-1700 μL

shipped in

ambient

General description

Millicell® EZ Slide allows the user to grow, fix, and stain cells directly on a microscope slide. There is no need to disassemble the unit or transfer cells from one device to another prior to analysis. The polypropylene wells are resistant to most fixatives used, so the cells can be grown on the slide and stained right in the well. Once stained, the microscope slide can be easily removed by pressing down the four side tabs of the unit and lifting the wells off of the slide. This easy slide removal reduces the risk of damaging the cells and no special tools are required to remove the wells from the slide. Raised corner ridges on the cover facilitate stacking of multiple units in the incubator, or they can be incubated in the tray supplied with the units.

Preparation Note

  1. Culture, fix, and stain the cells according to preferred protocol.
  2. To access the microscope slide, hold the Millicell® EZ SLIDE downon a flat surface and break the four tabs by pushing them downwith the thumb. Remove the cover. Hold the slide down and liftthe wells upward to remove. Remove slide from holder.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathilde Coustets et al.
Biomaterials, 241, 119908-119908 (2020-03-04)
The epithelial ovarian cancer is one of the most lethal gynecological malignancy due to its late diagnostic and many relapses observed after first line of treatment. Once diagnose, the most important prognostic factor is the completeness of cytoreductive surgery. To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service