Skip to Content
Merck

PC57

Anti-MSH2 (Ab-3) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Mismatch Repair Protein 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
human, mouse
Application:
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MSH2(4436)

General description

Purified rabbit polyclonal antibody. Recognizes the ~100 kDa MSH2 protein.
Recognizes the ~100 kDa MSH2 protein in HCT116 and SW480 cells and in colon tissue.
This Anti-MSH2 (Ab-3) Rabbit pAb is validated for use in Frozen Sections, Immunoblotting, Immunoprecipitation, Paraffin Sections for the detection of MSH2 (Ab-3).

Immunogen

full length, recombinant, human MSH2

Application

Frozen Sections (2 µg/ml)

Immunoblotting (1 µg/ml, chemiluminescence)

Immunoprecipitation (1 µg/sample)

Paraffin Sections (see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Other Notes

Bronner, C.E., et al. 1994. Nature368, 258.
Papadopoulos, N., et al. Science263, 1625.
Peltomäki, P.T. 1994. Ann. Medicine26, 215.
Fishel, R., et al. 1993. Cell75, 1027.
Leach, F.S., et al. 1993. Cell75, 1215.
Lindbolm, A., et al. 1993. Nat. Genet.5, 279.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Jennifer L Cyr et al.
Molecular carcinogenesis, 51(8), 647-658 (2011-08-13)
Lynch syndrome (LS) is caused by germline mutations in DNA mismatch repair (MMR) genes. MMR recognizes and repairs DNA mismatches and small insertion/deletion loops. Carriers of MMR gene variants have a high risk of developing colorectal, endometrial, ovarian, and other
J Barwell et al.
Journal of medical genetics, 44(8), 516-520 (2007-05-08)
Reports of differential mutagen sensitivity conferred by a defect in the mismatch repair (MMR) pathway are inconsistent in their conclusions. Previous studies have investigated cells established from immortalised human colorectal tumour lines or cells from animal models. We examined primary
Jennifer A McKinney et al.
Nature communications, 11(1), 236-236 (2020-01-15)
Alternative DNA structure-forming sequences can stimulate mutagenesis and are enriched at mutation hotspots in human cancer genomes, implicating them in disease etiology. However, the mechanisms involved are not well characterized. Here, we discover that Z-DNA is mutagenic in yeast as



Global Trade Item Number

SKUGTIN
PC57-100UG04055977207804