P2C100C05
Pellicon® 2 Cassette Ultrafiltration Module
filtration area 0.5 m2, NMWCO 100 kDa
Synonym(s):
Pellicon 2 Cassette Ultrafiltration Module PLCHK C 0.5 m2
Select a Size
CZK 97,800.00
Estimated to ship onJune 02, 2025Details
Select a Size
About This Item
CZK 97,800.00
Estimated to ship onJune 02, 2025Details
Recommended Products
material
composite regenerated cellulose (CRC)
polypropylene screen
polyurethane adhesive
Quality Level
manufacturer/tradename
Pellicon®
parameter
0.33 bar max. transmembrane pressure (5 psig) (reverse)
3.5 bar max. transmembrane pressure (50 psig) at 30 °C (forward)
5-35 L/min recirculation rate at 0.4 bar (6 psi)
technique(s)
analytical sample preparation: suitable
protein purification: suitable
H
1.5 cm (0.6 in.)
L
21 cm (8.3 in.)
W
17.8 cm (7.0 in.)
filtration area
0.5 m2
membrane nominal area
0.5 m2
gravimetric extractables
≤1500 mg/m2
matrix
Ultracel®
pore size
100 kDa NMWCO
pH
2-13
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Millipore′s PLCHK polyethersulfone membrane has void-free structures that guard against leakage of solutes through microdefects normally associated with voids beneath the thin skins of conventional UF membranes. These void-free membranes are more permeable, resulting in high-flux with equivalent or superior product retention. These void-free membranes provide the advantages of fast, high yield processing and smaller systems.
The Pellicon® 2 Cassette Ultrafiltration Module feature a filtration area of 0.5 m2 with a pore size of 100 kDa NMWCO.
Application
- Protein solutions
- Polysaccharide solutions
- Virus suspensions
Harvest, washing or clarification of:
- Cell cultures and lysates
- Colloidal suspensions
- Viral cultures
Features and Benefits
- Type C screen (coarse screen)
- Leading-edge void- free membranes to match virtually any separation challenge
- Short flow path for higher flux and higher resolution separation capability
- Choice of flow channel configuration providing process optimization capability
- Predictable, fast, scale-up
- True linear scalability from laboratory size modules to industrial assemblies for processing thousands of liters
- Brings higher resolution, improved yields and superior back-pressure resistance
- Ultracel membranes are composite membranes cast on a microporous substrate for defect-free membranes with superior
Other Notes
Analysis Note
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
A cost-model compares modalities like viral vectors and mRNA for production scale decision-making.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service