Skip to Content
Merck
All Photos(1)

Key Documents

OP14L

Sigma-Aldrich

Anti-c-ErbB2/c-Neu (Ab-2) Mouse mAb (9G6)

lyophilized, clone 9G6, Calbiochem®

Synonym(s):

Anti-HER2, Anti-Neu, Anti-ErbB2, Anti-Erythroblastosis Virus

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 9,080.00

CZK 9,080.00


Estimated to ship onMay 05, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 9,080.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 9,080.00


Estimated to ship onMay 05, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9G6, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ERBB2(2064)

General description

Anti-c-ErbB2/c-Neu (Ab-2), mouse monoclonal, clone 9G6, recognizes the ~185 kDa c-ErbB2/c-Neu SK-BR-3 cells and breast carcinoma. It is validated for IP and IHC on frozen and paraffin sections.
Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with SP2/0 Ag14 mouse myeloma cells. Recognizes the ~185 kDa c-ErbB2/c-neu protein.
Recognizes the ~185 kDa c-ErbB2/c-Neu in SK-BR-3 cells and breast carcinoma tissue. Does not cross-react with v-ErbB2 or the EGF receptor.

Immunogen

Human
cells overexpressing human c-ErbB2/c-Neu

Application

Frozen Sections (5 µg/ml)

Immunoprecipitation (1 µg/sample)

Paraffin Sections (see application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a volatile buffer, 100 µg BSA.

Reconstitution

Reconstitute the lyophilized antibody with sterile PBS, pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 µg/ml. Lyophilized antibodies should be reconstituted at 4°C with occasional gentle mixing for at least 2 h. Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage with 0.1% azide. Avoid freeze/thaw cycles of solutions.

Analysis Note

Negative Control
HepG2 cells or normal skin
Positive Control
SK-BR-3 cells or breast carcinoma tissue

Other Notes

Immunoprecipitates the ~185 kDa c-neu protein and has been reported to stain formalin-fixed paraffin-embedded tissue sections of human breast carcinomas overexpressing the c-neu protein. This antibody reacts with a cell surface epitope of c-neu but does not cross-react with the EGF receptor. In one study ductal carcinomas in situ displaying large-cell, comedo growth type, were stained while none of 16 ductal carcinomas in situ of small-cell, papillary, or cribiform growth type were stained by this antibody. Antibody should be titrated for optimal results in individual systems.
Van De Vijver, M.J., et al.1988 New Engl. J. Med.319, 1239.
DiFiore, P.P., et al.1987 Science237, 178.
Slamon, D.J., et al. 1987 Science235, 177.
Varley, J.M., et al.1987 Oncogene1, 423.
Bargmann, C.I., et al. 1986 Nature319, 226.
Yamamoto, T., et al.1986 Nature319, 230.
Blick, M., et al. 1984 Blood64, 1234.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service