Skip to Content
Merck

Skip To

NA02

Anti-2,2,7-Trimethylguanosine Mouse mAb (K121)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

12 500,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Species reactivity:
(human, mouse, rat)
Species reactivity:
-
Application:
Citations:
-

12 500,00 CZK


Estimated to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

form

liquid

species reactivity

(human, mouse, rat)

storage condition

do not freeze

dilution

(Affinity Purification
Immunocytochemistry (0.5-5 µg/mL)
Immunoprecipitation (1-2 µg)
Paraffin Sections )

isotype

IgG1

storage temp.

2-8°C

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells. Recognizes the exposed 5′ cap structure (2,2,7-trimethylguanosine) of small nuclear RNAs (snRNAs).

Immunogen

2,2,7-trimethylguanosine

Application

Affinity PurificationImmunocytochemistry (0.5-5 µg/mL)Immunoprecipitation (1-2 µg) Paraffin Sections


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library