Skip to Content
Merck
All Photos(1)

Key Documents

MXQREAG48

Millipore

Milliflex® Quantum reagents

pkg of 48 tests, suitable for bioburden testing

Synonym(s):

Fluorescence-based rapid detection system

Sign Into View Organizational & Contract Pricing

Select a Size

48 PKG
CZK 6,460.00

CZK 6,460.00


In StockDetails


Request more information

Select a Size

Change View
48 PKG
CZK 6,460.00

About This Item

UNSPSC Code:
41116130
NACRES:
NB.79

CZK 6,460.00


In StockDetails


Request more information

packaging

pkg of 48 tests

Quality Level

manufacturer/tradename

Milliflex® Quantum

application(s)

bioburden testing

storage temp.

2-8°C

General description

Milliflex® Quantum Reagents are a part of Milliflex® Quantum system which is a convenient and sensitive platform for the rapid quantitative detection of contaminants in filterable samples. This rapid microbiological method is based on a universal enzymatic fluorescent staining of viable and culturable microorganisms. The fluorescent staining procedure is non-destructive and allows for identification of microorganisms after a positive result. Therefore, the Milliflex® Quantum system is a fast and reliable alternative for the detection of microorganisms.

Application

The Milliflex® Quantum Reagents are used for rapid bioburden testing in filtrable samples, including in-process and final product samples (pharmaceuticals and water). This product is also used in industrial QC and water monitoring.

Features and Benefits

  • Non-destructive method enables reliable identification using any ID technology
  • Easy-to-use system with simple workflow requires minimal training
  • Results comparable to compendial method, enabling faster validation, benefit of rapid detection
  • Economical, robust systemCompact hardware fits on any laboratory bench

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service