Skip to Content
Merck
All Photos(1)

Documents

MR-122

Sigma-Aldrich

EmbryoMax® FHM Mouse Embryo Media

(1X), Liquid, w/o Phenol Red and BSA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.71

Quality Level

form

liquid

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | embryo: suitable
cell culture | stem cell: suitable

input

sample type: mouse embryo(s)

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service