Skip to Content
Merck

Skip To

MPXPVER-K25

MAGPIX Performance Verification Kit, 25 uses

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 each

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

13 400,00 CZK

About This Item

UNSPSC Code:
41116010
NACRES:
HB.01
eCl@ss:
42029053
Technique(s):
multiplexing: suitable

13 400,00 CZK


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Milliplex®

technique(s)

multiplexing: suitable

General description

All xMAP Instruments require regular calibration and performance verification testing, to ensure the system is operating correctly and to maintain data accuracy. The MAGPIX instrument uses a calibration kit and a verification kit, each providing 25 uses.

Application

For use with MAGPIX® system

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Legal Information

MAGPIX is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters