Skip to Content
Merck
All Photos(1)

Key Documents

MPE030025

Millipore

Microcon® Centrifugal Filters

MWCO 30 kDa, polyethersulfone membrane (Biomax), sample volume 0.5 mL

Synonym(s):

Biomax polyethersulfone (PES) membrane centrifuge filter

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
CZK 3,320.00

CZK 3,320.00


Available to ship onApril 10, 2025Details



Select a Size

Change View
25 EA
CZK 3,320.00

About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

CZK 3,320.00


Available to ship onApril 10, 2025Details


material

polyacetal base (membrane support)
polycarbonate device top
polyethersulfone membrane (Biomax)
polypropylene tube (for filtrate/concentrate)
silicone O-ring (medical grade)

sterility

non-sterile

product line

Microcon®

feature

holdup volume> 10 μL

parameter

0.5 mL sample volume

D

12.3 mm

L

4.5 cm (1.8 in.)

filtration area

0.32 cm2

pore size

30 kDa MWCO

Application

These 0.5 mL centrifugal filter enables the concentration of biomolecules with a NMWL of 30kDa. Microcon® centrifugal filters are ideal for concentration & desalting.

Features and Benefits

  • High recovery for small volumes with reverse spin (concentration factor less than 20X)
  • Biomax membrane
  • Fast processing

Physical form

Color Code: Pink

Legal Information

MICROCON is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service