Skip to Content
Merck

Skip To

MB000000R

Rosolic Acid (Dehydrated)

Selective agent to be added to mFC broth medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea

Estimated to ship onMay 29, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 6,410.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74
eCl@ss:
32160801

CZK 6,410.00


Estimated to ship onMay 29, 2026Details


Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder (dehydrated)

shelf life

12 mo.

packaging

pkg of 25 g

color

yellow to red

input

sample type bottled water
sample type soft drink
sample type cosmetics
sample type sport drink
sample type cider
sample type wine
sample type food(s)
sample type beverage(s)
sample type beer

pH

6.8±8.2 (25.0 °C)

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc
microbiology
water monitoring

compatibility

for use with MF-method, for use with Microfil®, for use with MicropreSure®, for use with Milliflex®

shipped in

ambient

storage temp.

2-30°C

Application

Selective agent to be added to mFC broth medium
Type of Test
Bioburden

Microbial Limits

Features and Benefits

Incubation Time: 24 hrs ± 2

Packaging

Dehydrated 25 g in plastic bottle

Preparation Note

Rosolic Acid

Analysis Note

Target Microorganisms: Rinse Buffers and Additives

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MB000000FMB000000TMB000000E
form

powder (dehydrated)

form

powder (dehydrated)

form

powder (dehydrated)

form

powder (dehydrated)

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc
microbiology
water monitoring

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc (microbial monitoring)
microbiology
water monitoring

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc
water monitoring, microbiology

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc
water monitoring, microbiology

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

compatibility

for use with MF-method, for use with MicropreSure®, for use with Microfil®, for use with Milliflex®

compatibility

for use with MF-method, for use with Microfil®, for use with MicropreSure®, for use with Milliflex®

compatibility

for use with MF-method, for use with Microfil®, for use with MicropreSure®, for use with Milliflex®

compatibility

for use with MF-method, for use with Microfil®, for use with MicropreSure®, for use with Milliflex®

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

pH

6.8±8.2 (25.0 °C)

pH

7.4±0.2 (25.0 °C)

pH

7.0±0.2 (25.0 °C)

pH

7.2±0.1 (25.0 °C)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters