Merck
MANTA4KIT
MAS-100 NT® Validation Protocol and Test Record Protocol Kit
A4 format
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.79
UNSPSC Code:
41116500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
compatibility
for use with MAS-100 NT® air sampler system.
General description
MAS-100 NT® validation protocol and associated test record protocol kit is designed as a ready to use guide to perform the validation activities for the MAS-100 NT® air sampling pump. The validation protocol kit in A4 format ensures regulatory compliance of the pumps and guarantees consistent performance.
Application
MAS-100 NT® validation protocol kit is used for the validation of MAS-100 NT® pumps which samples environmental air inside cleanrooms and isolators.
Legal Information
MAS-100 NT is a registered trademark of MBV AG