Skip to Content
Merck

Skip To

MANMN4010

Multiscreen® 96 well Plate, mesh nylon membrane

pore size 40 μm, non-sterile

Synonym(s):

Mesh Nylon 96-Well Plate, Nylon Membrane Plate, 96 microtiter plates, 96 multiwell plates, 96 well filter plates, 96 well microplates, 96 well plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 ea

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

20 400,00 CZK

About This Item

UNSPSC Code:
41104923
NACRES:
NB.22
eCl@ss:
32039006
Material:
clear polystyrene plate, flat bottom wells, mesh Nylon membrane
Pore size:
40 μm pore size
Sterility:
non-sterile

20 400,00 CZK


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

clear polystyrene plate, flat bottom wells, mesh Nylon membrane

description

Whole Organism Screening

sterility

non-sterile

usage

Manual

feature

hydrophilic, lid

packaging

pkg of 10 plates

manufacturer/tradename

MultiScreen®

technique(s)

Particulate-laden sample filtration: suitable, prefiltration: suitable

filtration area

0.28 cm2

plate size

96 wells

well maximum volume

300 μL

working volume

50-250 μL

pore size

40 μm pore size

application(s)

sample preparation

compatibility

manifold only

shipped in

ambient

storage temp.

no temp limit

General description

MultiScreen-Mesh plate with receiver plate provides a complete system for culturing, testing and reading assay results in a one-step protocol. This system is ideal for target screening and other applications that evaluate new compounds using multicellular organisms as the in vivo model.

MultiScreen-Mesh is optimized for compound and target identification in a 96-well format. The 96-well nylon mesh plate and 96-well tray fit together to provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms. Select from a range of mesh pore sizes to appropriate for your application. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms. The plates are optically clear for use with an imaging microscope or other microscopic analysis. Plates can be coated with a thin layer of agar if necessary.

MultiScreen-Mesh plates have also been shown to be useful as high throughput cell strainers prior to cellular analysis in a FACS analyzer. To dissociate clumps of cells prior to loading plate into FACS instrument, run cell suspensions through the mesh plates either by gravity flow or via gentle centrifugation.

This filter plate provides a complete system for culturing, testing and reading assay results in a one-step protocol. The system is ideal for target screening and other applications that evaluate the effect of new compounds on whole, multicellular organisms. The MultiScreen-Mesh filter plate is optimized for compound and target identification in a 96-well format. The filter plate and 96-well tray fit together to provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms.

Features & Benefits:
- Multiple configurations and applications
- Chemical compatibility
- Automation-compatible
- Superior vacuum filtration and filtrate collection
- Plates are optically clear for use with an imaging microscope or other microscopic analysis

Applications
- Whole Organism Screening

Application

Research Category
Whole Organism
Whole Organism Screening

Physical form

Color Code: Clear

Other Notes

Automation Compatible: No

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MANMN2010MANMN6010MANM10010
pore size

40 μm pore size

pore size

20 μm pore size

pore size

60 μm pore size

pore size

100 μm pore size

manufacturer/tradename

MultiScreen®

manufacturer/tradename

MultiScreen®

manufacturer/tradename

MultiScreen®

manufacturer/tradename

MultiScreen®

material

clear polystyrene plate, mesh Nylon membrane, flat bottom wells

material

clear polystyrene plate, flat bottom wells, mesh Nylon membrane

material

clear polystyrene plate, flat bottom wells, mesh Nylon membrane

material

clear polystyrene plate, flat bottom wells, mesh Nylon membrane

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

well maximum volume

300 μL

well maximum volume

300 μL

well maximum volume

300 μL

well maximum volume

300 μL

feature

hydrophilic, lid

feature

hydrophilic, lid

feature

hydrophilic, lid

feature

hydrophilic, lid


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters