Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
10 ea | Estimated to ship on18 August 2026fromAreál Kühne+Nagel spol. s r.o. | 27 100,00 CZK |
About This Item
27 100,00 CZK
Estimated to ship on18 August 2026Details
Quality Segment
material
clear polystyrene plate, flat bottom wells, polycarbonate membrane
description
Clear, Sterile, 3.0 µm pore size, Polycarbonate (PC) membrane, 10 plates
sterility
sterile
product line
MultiScreen®
usage
Manual
feature
hydrophilic, lid
packaging
pkg of 10 plates
technique(s)
DNA purification: suitable, cell based assay: suitable, cell migration: suitable
filtration area
0.28 cm2
plate size
96 wells
well maximum volume
350 μL
working volume
50-250 μL
pore size
3 μm pore size
binding type
Tissue Culture (TC)-treated surface
compatibility
manifold only
shipped in
ambient
storage temp.
10-30°C
General description
HTS Filter Plates (Plate catalogue numbers that start with MS)
- MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.
- Millipore’s new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.
Classic Filter Plates (Plate catalogue numbers that start with MA)
- The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.
Accessory – MultiScreenHTS Vacuum Manifold
- The MultiScreenHTS Vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.
Millicell Assay Plates
- In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.
Application
Nucleic acid
Physical form
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description pore size 3 μm, sterile | description pore size 8 μm, sterile | description pore size 0.4 μm, non-sterile | description pore size 5 μm, sterile |
| sterility sterile | sterility sterile | sterility non-sterile | sterility sterile |
| manufacturer/tradename MultiScreen® | manufacturer/tradename MultiScreen® | manufacturer/tradename MultiScreen® | manufacturer/tradename MultiScreen® |
| binding type Tissue Culture (TC)-treated surface | binding type Tissue Culture (TC)-treated surface | binding type Tissue Culture (TC)-treated surface | binding type Tissue Culture (TC)-treated surface |
| pore size 3 μm pore size | pore size 8 μm pore size | pore size 0.4 μm pore size | pore size 5 μm pore size |
| material clear polystyrene plate, polycarbonate membrane, flat bottom wells | material clear polystyrene plate, flat bottom wells, polycarbonate membrane | material clear polystyrene plate, flat bottom wells, polycarbonate membrane | material clear polystyrene plate, flat bottom wells, polycarbonate membrane |
| well maximum volume 350 μL | well maximum volume 350 μL | well maximum volume 300 μL | well maximum volume 350 μL |
Still not finding the right product?
Explore all of our products under Multiscreen® 96 well Plate, polycarbonate membrane
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

